Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Chemical and Hazardous Materials Signs: DANGER - FLAMMABLE MATERIALS

Catalog No. 19000659 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Aluminum
Fiberglass
Plastic
Polyester
Height (Metric):
25.4 cm
25.40 cm
Width (Metric):
25.40 cm
35.5 cm
35.56 cm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (Metric) Width (Metric)
19-000-659 Aluminum 25.4 cm 35.5 cm
17-046-22 Fiberglass 25.40 cm 35.56 cm
19-017-216 Polyester 25.40 cm 35.56 cm
19-035964 Plastic 25.40 cm 35.56 cm
19-035965 Polyester 25.40 cm 25.40 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-000-659 Supplier Brady™ Supplier No. 43244
Add to Cart
Add to Cart

Helps identify plant's chemical storage and hazardous materials areas

  • Available in three types of materials: fiberglass, self-sticking polyester and plastic
  • Available in various widths and heights
  • Black/red on white

Specifications

Certifications/Compliance ISO 9001, ANSI Z535, OSHA 1910.145
Material Aluminum
For Use With (Application) Designed for use in industrial, utility, commercial and institutional environments
Height (English) 10 in.
Height (Metric) 25.4 cm
Width (English) 14 in.
Width (Metric) 35.5 cm
Displayed Text Flammable Material
Product Type Hazardous Materials Sign
Color Black

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.