Learn More
Brady™ Code Reader™ CR1400 Barcode Readers
Catalog No. 19104464 Shop All Brady Corporation Products
Change view
Click to view available options
Includes:
1.8 m USB Cable and Stand
Catalog No. Includes
19-104-464 1.8 m USB Cable and Stand
1 options
Catalog No. 19-104-464 Supplier Brady™ Supplier No. 133070
Used for data collection and tracking applications
  • Compact, rugged
  • Features high performance scan engine and intuitive targeting
  • High speed, omnidirectional reading of 1D and 2D barcodes
  • Dual field optics, both high density and wide field in same unit
  • Glare reduction technology for reading codes on shiny surfaces
  • Manual or automatic triggering
  • 3 programmable indicators: vibration mode, LED and tone
  • Data editing and parsing with JavaScript
  • Disinfectant ready, IP54 housing
Specifications
Includes 1.8 m USB Cable and Stand
Type Barcode Reader