Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Compact Keyed Alike 1 in. Shackle Aluminum Padlocks

Catalog No. 19103536 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
Silver
Yellow
Quantity:
3/Pk.
6/Pk.
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
19-103-536 Yellow 3/Pk.
19-103-533 Red 3/Pk.
19-103-542 Red 6/Pk.
19-103-543 Blue 6/Pk.
19-103-544 Green 6/Pk.
19-103-545 Yellow 6/Pk.
19-103-546 Orange 6/Pk.
19-103-547 Black 6/Pk.
19-103-548 Brown 6/Pk.
19-103-549 Purple 6/Pk.
19-103-550 Silver 6/Pk.
19-103-534 Blue 3/Pk.
19-103-535 Green 3/Pk.
19-103-537 Orange 3/Pk.
19-103-538 Black 3/Pk.
19-103-539 Brown 3/Pk.
19-103-540 Purple 3/Pk.
19-103-541 Silver 3/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 19-103-536 Supplier Brady™ Supplier No. 133282

Compact Keyed Alike 1 in. Shackle Aluminum Padlocks include two keys per lock.

  • Compact lock body measures 1-1/4 in. H x 1-1/4 in. W
  • Bright, anodized aluminum finish provides greater visibility and fade-resistance in all environoments
  • Reserved, paracentric keyway provides optimal security (prevents someone from getting a copy made at a local key shop)
  • Every lock in this set can be opened with the same key, which is beneficial when multiple locks are assigned to a single employee

Specifications

Certifications/Compliance OSHA 1910.147
Color Yellow
Key Type Keyed Alike
Material Aluminum
Quantity 3/Pk.
Product Type Padlock
For Use With (Application) Outdoor; Industrial Manufacturing; Food and Beverage; Confined Spaces
For Use With (Equipment) Outdoor; Industrial Manufacturing; Food and Beverage; Confined Spaces
Width (English) Shackle 1 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.