Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Compact Keyed-Different 1 in. Shackle Aluminum Padlocks

Catalog No. 19103519 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
Silver
Yellow
Quantity:
1 Ea.
6/Pk.
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
19-103-519 Orange 6/Pk.
19-103-517 Green 6/Pk.
19-103-520 Black 6/Pk.
19-103-524 Red 1 Ea.
19-103-525 Blue 1 Ea.
19-103-526 Green 1 Ea.
19-103-527 Yellow 1 Ea.
19-103-528 Orange 1 Ea.
19-103-529 Black 1 Ea.
19-103-530 Brown 1 Ea.
19-103-531 Purple 1 Ea.
19-103-532 Silver 1 Ea.
19-103-515 Red 6/Pk.
19-103-516 Blue 6/Pk.
19-103-518 Yellow 6/Pk.
19-103-521 Brown 6/Pk.
19-103-522 Purple 6/Pk.
19-103-523 Silver 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 19-103-519 Supplier Brady™ Supplier No. 133265

Compact Keyed-Different 1 in. Shackle Aluminum Padlocks include two keys per lock.

  • Compact lock body measures 1-1/4 in. H x 1-1/4 in. W
  • Bright, anodized aluminum finish provides greater visibility and fade-resistance in all environoments
  • Reserved, paracentric keyway provides optimal security (prevents someone from getting a copy made at a local key shop)
  • Each padlock is keyed differently with keys precision-machined to exacting standards, so that the key for one lock will not open another in the set

Specifications

Certifications/Compliance OSHA 1910.147
Color Orange
Key Type Keyed Different
Material Aluminum
Quantity 6/Pk.
Product Type Padlock
For Use With (Application) Outdoor; Industrial Manufacturing; Food and Beverage; Confined Spaces
For Use With (Equipment) Outdoor; Industrial Manufacturing; Food and Beverage; Confined Spaces
Width (English) Shackle 1 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.