Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Danger: Benzene Signs

Catalog No. 19035697 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Plastic
Polyester
Temperature Range (English):
-40°F to 175°F
-40°F to 230°F
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Temperature Range (English)
19-035-697 Plastic -40°F to 175°F
19-035-699 Polyester -40°F to 230°F
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-035-697 Supplier Brady™ Supplier No. 22723

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads “DANGER BENZENE”
  • Black/red on white
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

Specifications

Material Plastic
Quantity 1 Ea.
For Use With (Application) Light duty industrial, utility, commercial, institutional environments
Height (English) 7 in.
Height (Metric) 17.7 cm
Max. Temperature (English) 175°F
Min. Temperature (English) -40°F
Thickness 0.06 in.
Width (English) 10 in.
Width (Metric) 25.4 cm
Temperature Range (Metric) -40°C to 80°C
Temperature Range (English) -40°F to 175°F
Dimensions (L x W) 10 x 7 in.
Displayed Text DANGER Benzene
Product Type Chemical and Hazardous Materials Sign
Color Red on White
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.