Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Danger: Chemical Waste Storage Area… Signs

Catalog No. 19035583 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Plastic
Polyester
Height (English):
10 in.
10 in.10 in.
7 in.
Width (English):
10 in.
14 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (English) Width (English)
19-035583 Polyester 7 in. 10 in.
19-035-582 Plastic 10 in. 14 in.
19-035584 Polyester 10 in.10 in. 14 in.
19-035581 Plastic 7 in. 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-035583 Supplier Brady™ Supplier No. 84042

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads “DANGER CHEMICAL WASTE STORAGE AREA UNAUTHORIZED PERSONS KEEP OUT”
  • Black/red on white
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene
  • B-302
    • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic

    Specifications

    Certifications/Compliance ANSI Z535, OSHA 1910.145, ISO 9000
    Material Polyester
    Quantity 1 Ea.
    For Use With (Application) Identification of fire alarms, extinguishers and emergency exits
    Height (English) 7 in.
    Height (Metric) 17.7 cm
    Max. Temperature (English) 230°F
    Min. Temperature (English) -40°F
    Thickness 0.006 to 0.01 in.
    Width (English) 10 in.
    Width (Metric) 25.4 cm
    Temperature Range (Metric) -40°C to 110°C
    Temperature Range (English) -40°F to 230°F
    Displayed Text DANGER Chemical Waste Storage Area Unauthorized Persons Keep Out
    Product Type Admittance Sign
    Color Red on White
    Show More Show Less

    Adhere to
    Safety Regulations and Best Practices

    Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.