Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Danger: Highly Combustible Area... Signs

Catalog No. 19035977 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Plastic
Polyester
Height (English):
7 in.
10 in.
Width (English):
10 in.
14 in.
Thickness:
0.006 to 0.01 in.
0.06 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (English) Width (English) Thickness
19-035-977 Plastic 10 in. 14 in. 0.06 in.
19-035-976 Plastic 7 in. 10 in. 0.06 in.
19-035-979 Polyester 10 in. 14 in. 0.006 to 0.01 in.
19-035-978 Polyester 7 in. 10 in. 0.006 to 0.01 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-035-977 Supplier Brady™ Supplier No. 25675

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical and hazardous materials Signs that identify equipment, rooms and materials which contain or are contaminated with hazardous agents

  • Signs read as “DANGER - HIGHLY COMBUSTIBLE AREA NO WELDING, BURNING OR OPEN FLAMES PERMITTED”
  • Available in three materials - plastic, fiberglass and polyester
  • Available in various widths and heights
  • Black/red on white

Specifications

Certifications/Compliance ANSI Z535, OSHA 1910.145
Material Plastic
Quantity 1 Ea.
Mounting Type Surface Mount
For Use With (Application) Designed for use in industrial, utility, commercial and institutional environments
Height (English) 10 in.
Height (Metric) 25.40 cm
Max. Temperature (English) 175°F
Min. Temperature (English) -40°F
Thickness 0.06 in.
Width (English) 14 in.
Width (Metric) 35.56 cm
Temperature Range (Metric) -40°C to 80°C
Temperature Range (English) -40°F to 175°F
Displayed Text Highly Combustible Area No Welding, Burning or Open Flame Permitted
Product Type Hazardous Materials Sign
Color Red on White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.