Description
- The Electrical Lockout Toolbox Kit from Brady™ gives you all the basics to lockout breakers, electrical plugs, wall switches and fuses
- Comes in an easy to transport extra-large toolbox and also contains a red belt pouch
- Equipping your employees with the proper lockout tools and warning devices can save lives, reduce lost employee time, and cut insurance costs
- Brady™ is a world leader in lockout/tagout products and services offering industry expertise and a broad range of durable, easy-to-use lockout devices
Specifications
Specifications
| Product Type | Electrical Lockout Toolbox Kit |
| Certifications/Compliance | OSHA 1910.147 |
| Depth (English) | 8 in. |
| Includes | (1) Mini Cable Lockout, (2) 120V Snap-On Breaker Lockouts, (2) 120/277V Clamp-On Breaker Lockouts, (2) 480/600V Clamp-On Breaker Lockouts, (2) Universal Multi-Pole Breaker Lockouts, (1) Oversized Breaker Lockout, (2) Cleats for 120/277V Clamp-On Lockout, (2) Cleats for 480/600V Clamp-On Lockout, (1) 3-In-1 Plug Lockout, (1) Wall Switch Lockout, (1) Small Fuse Blockout, (1) Large Fuse Blockout and (1) Extra-Large Lockout Toolbox, (1) Red Belt Pouch |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.