Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ ENV™ - MAXX Enhanced Oil Only Sorbent Pads

Catalog No. 17111141 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
24.5 gal./Bale
28 gal./Bale
33 gal./Bale
50 gal./Bale
51 gal./Bale
Weight Capacity:
Heavyweight
Lightweight
Medium-weight
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Weight Capacity
17-111-141 50 gal./Bale Medium-weight
19-000-161 24.5 gal./Bale Lightweight
17-111-135 51 gal./Bale Lightweight
19-000-159 33 gal./Bale Heavyweight
19-000-160 28 gal./Bale Medium-weight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-111-141 Supplier Brady™ Supplier No. ENV50
Add to Cart
Add to Cart

Oil only sorbent equally effective on land or water

  • Single-ply construction
  • Repels water and floats indefinitely even when saturated
  • For use with oil and petroleum-based fluids
  • Available in two sizes
  • MAXX refers to enhanced performance without extra weight

    Applications:

    Marine applications, over spray leaks and drips, workstations, drums - liquid storage, outfalls and discharge ponds, emergency spill response.

    Specifications

    Color White
    Capacity (English) 50 gal./Bale
    Material Polypropylene
    Type Absorbent Pad, Oil-only
    For Use With (Application) Overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge ponds
    Packaging Quantity 50/Cs.
    Weight Capacity Medium-weight
    Dimensions (L x W) 76.2 x 76.2 cm (30 x 30 in.)
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.