Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Floor Safety Signs

Catalog No. 19054841 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black on White
Black on Yellow
Green on White
Purple on White
Red on White
White on Red
Displayed Text:
Ear Protection Area
Emergency Exit
Emergency Stairs
Eye Wash
Fire Extinguisher
Fire Hose
Hard Hat Area
Keep Aisles Clear
No Smoking
Pedestrian Traffic Only
Safety Glasses Required
Safety Goggles Required
Stop
Watch Out For Forklift Trucks
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Displayed Text
19-054-841 Red on White Emergency Stairs
19-054-837 Red on White Fire Extinguisher
19-054-840 Red on White Emergency Exit
19-054-889 Black on Yellow Ear Protection Area
19-054-891 Black on Yellow Hard Hat Area
19-054-892 Black on Yellow Safety Goggles Required
19-054-897 Green on White Eye Wash
19-054-899 Black on White Pedestrian Traffic Only
19-054-898 Purple on White Keep Aisles Clear
19-054-900 Black on Yellow Safety Glasses Required
19-054-901 White on Red Stop
19-054-839 White on Red Fire Hose
19-054-887 Red on White No Smoking
19-054-888 Black on Yellow Watch Out For Forklift Trucks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 19-054-841 Supplier Brady™ Supplier No. 49068

Durable, highly visible Floor Safety Signs are designed to warn of hazardous areas, putting valuable safety information at the point of need.

  • Floor safety signs improve facility safety by alerting employees to hazards and/or important safety information in areas with foot traffic
  • B-819 Anti-Skid Vinyl features a velvet embossed rigid overlaminate
  • Excellent chemical, oil and water resistance
  • Minimum application temperature of 0°F (-18°C)

Specifications

Certifications/Compliance ASTM 4.2.1
Material Vinyl
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Diameter (English) 17 in.
Mounting Type Surface Mount
For Use With (Application) Designed to warn of hazardous areas, putting valuable safety information at the point of need
Max. Temperature (English) 158°F
Min. Temperature (English) -40°F
Thickness 0.01 in.
Temperature Range (Metric) -40°C to 70°C
Temperature Range (English) -40°F to 158°F
Displayed Text Emergency Stairs
Product Type Floor Safety Sign
Color Red on White
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.