Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ GHS Wallet Cards

Catalog No. 19141907 Shop All Brady Corporation Products
Change view
Click to view available options
Width (Metric):
5.3 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
19-141-907 5.3 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-141-907 Supplier Brady™ Supplier No. 133208
This item is not returnable. View return policy

Outlines all 9 of the new GHS Hazard Pictograms

  • Training aid to help employees get familiar with new hazard pictograms
  • Handy reference for when new GHS labels are entering facility
  • 10 cards per pack
  • Dimensions: 2.1W x 3.4 in. H

Specifications

Includes Outlines all 9 of new GHS hazard pictograms
Quantity 10/Pk.
For Use With (Application) Training aid to help employees get familiar with new hazard pictograms
Height (English) 3.4 in.
Height (Metric) 8.6 cm
Width (English) 2.1 in.
Width (Metric) 5.3 cm
Product Type GHS Wallet Card

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.