Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ GlobalMark™ Label Printer: Hard Carrying Case with Wheels

Catalog No. 19201241 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-201-241 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-201-241 Supplier Brady™ Supplier No. 76802

GlobalMark™ Label Printer: Hard Carrying Case with Wheels protects your GlobalMark™ Series sign and label printer and keeps accessories organized with a hard case.

  • Package includes the Printer Case only (printer sold separately)
  • Designed for use with the GlobalMark™ Series Sign and Label Printers from Brady™
  • The durable hard case protects your printer against impacts and dust while allowing for easy mobility
  • Keeps printer accessories organized and on hand

Specifications

Product Type Hard Case
Color Black
For Use With (Equipment) GlobalMark Printer
Sterility Non-sterile
Format Ribbon
Includes Wheels
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.