Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ GlobalMark™ Single-Color Printer Ribbons

Catalog No. 19104564 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Cyan
Green
Magenta
Orange
Purple
Red
White
Yellow
Yellow/Transparent
For Use With (Application):
Indoor and outdoor use
Process Colors
Right-to-Know Labels
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color For Use With (Application)
19-104-564 Orange Indoor and outdoor use
19-201-216 Black Indoor and outdoor use
19-201-218 Red Indoor and outdoor use
19-201-219 Blue Indoor and outdoor use
19-201-217 White Indoor and outdoor use
19-104-561 Green Indoor and outdoor use
19-104-563 Yellow Indoor and outdoor use
19-104-572 Black Right-to-Know Labels
19-104-562 Magenta Indoor and outdoor use
19-104-565 Cyan Process Colors
19-104-566 Yellow/Transparent Indoor and outdoor use
19-104-567 Purple Indoor and outdoor use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 19-104-564 Supplier Brady™ Supplier No. 76752
Add to Cart
Add to Cart

Exceptionally tough, smear-proof ribbons; printed text lasts years outdoors and in industrial environments

  • Self-contained ribbon cartridges drop straight into GlobalMark™ label makers; click into place

Order Info

Dimensions (W x L): 4.11 in. x 200 ft. (10.4cm x 61m). Two- and four-color ribbons also available; contact your Fisher Scientific Customer Service Representative.

Specifications

Product Type Printer Ribbon
Color Orange
For Use With (Equipment) GlobalMark Label Maker
Length (English) 200 ft.
Width (English) 3.25 in.
Length (Metric) 60.96 m
Format Cartridge, Ribbon
Certifications/Compliance RoHS 2005/618/EC MCV, 2002/95/EC
For Use With (Application) Indoor and outdoor use
Quantity 1 Ea.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.