Learn More
Description
- The B-928 is a flexible vinyl film with a cold temperature applicable pressure sensitive adhesive
- Service temperature range of -50 to 176°F
- Average outdoor durability of 2 years
- RoHS Compatibility Pass 2011/65/EU
Specifications
Specifications
| Certifications/Compliance | RoHS |
| Adhesive Type | Acrylic, Pressure-sensitive |
| Color | Black, Yellow |
| Format | Label |
| Material | Vinyl |
| Product Type | Hazardous Waste Label |
| Sterility | Non-sterile |
| Display Text | Acetone |
| For Use With (Application) | Hazardous waste material labels |
| Gloss | 6 Gardner Units |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.