Learn More
Description
- Chemically inert — will not react with aggressive fluids
- Color-coded green indicates use with hazardous materials
- Great for absorbing spills and leaking fluids in tight spaces
- Class: chemical
HAZ1818
- Shipping weight: 8.62kg (19 lbs.); 16 per case
- Shipping weight: 5.44kg (12 lbs.); 32 per case
Specifications
Specifications
| Color | Green |
| Capacity (English) | 21 gal. |
| Capacity (Metric) | 79.38 L |
| Material | Polypropylene |
| Type | Hazwik Sorbent Pillow |
| For Use With (Application) | Provide extra protection during packaging and transport, absorbs chemicals, acids and bases, spills and leaking fluids in tight spaces |
| Packaging Quantity | 32/Cs. |
| Dimensions (L x W) | 9 x 9 in. |
| Weight (English) | 12 lb. |
| Weight (Metric) | 5.44 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.