Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Heavy Duty Anti-Skid Tapes

Catalog No. 19103675 Shop All Brady Corporation Products
Change view
Click to view available options
Width (English):
2 in.
4 in.
6 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
19-103-675 4 in.
19-103-649 6 in.
19-103-676 2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-103-675 Supplier Brady™ Supplier No. 110019
Add to Cart
Add to Cart

For non-slip protection on hazardous surfaces

  • Durable, high slip resistance with deeply textured silicon carbide grit
  • Provides high traction surface even in dirtiest work environments
  • 46-grit non-skid offers longevity in both indoor and outdoor applications
  • Acrylic adhesive system allows adhesion over a wide variety of surfaces, while offering water and chemical resistance
  • Engineered to withstand harsh elements such as oil, fuel and grease often found in the most extreme MRO/OEM environments

Applications:

  • Loading docks, ramps

Specifications

Color Black
Certifications/Compliance OSHA Standards, NSFI - National Flooring Safety Institute certified for high traction
Length (Metric) 18.2 m
Width (English) 4 in.
Width (Metric) 10.1 cm
Adhesive Type Acrylic
Type Anti-Skid Tape
Length (English) 60 ft.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.