Learn More
Brady™ Key Retaining Keyed Alike Steel Padlocks with 3 inch Shackle Locks
Shop All Brady Corporation ProductsDescription
Includes
1 key
- Key-retaining steel padlock has a 3 in. reinforced laminated steel shackle
- Keyed alike
- Hardened steel body withstands severe physical abuse and tampering
- Offers individually coated body plates that provide superior rust protection and corrosion resistance
- Available in a variety of different colors
Specifications
Specifications
| Color | Purple |
| Quantity | 3/Pk. |
Safety and Handling
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
