Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Key Retaining Keyed Different Steel Padlocks with 2 inch Shackle Locks

Catalog No. 19103417 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Brown
Purple
Quantity:
1 Ea.
6/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
19-103-417 Brown 1 Ea.
19-103-418 Purple 1 Ea.
19-103-425 Brown 6/Pk.
19-103-426 Purple 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-103-417 missing translation for 'mfr' Brady™ missing translation for 'supplierNo' 123274

Includes

1 key

Key retaining safety feature prevents key from being removed until closed

  • Key-retaining steel padlock has a 2 in. reinforced laminated steel shackle
  • Hardened steel body withstands severe physical abuse and tampering
  • Offers individually coated body plates that provide superior rust protection and corrosion resistance
  • Available in a variety of different colors

Specifications

Color Brown
Quantity 1 Ea.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.