Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Key Retaining Keyed Different Steel Padlocks with 3 inch Shackle Locks

Catalog No. 19103393 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Purple
Red
White
Yellow
Quantity:
1 Ea.
6/Pk.
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
19-103-393 Blue 1 Ea.
19-103-383 Red 6/Pk.
19-103-384 Green 6/Pk.
19-103-385 Blue 6/Pk.
19-103-387 White 6/Pk.
19-103-386 Yellow 6/Pk.
19-103-388 Black 6/Pk.
19-103-389 Brown 6/Pk.
19-103-390 Purple 6/Pk.
19-103-391 Red 1 Ea.
19-103-392 Green 1 Ea.
19-103-395 White 1 Ea.
19-103-394 Yellow 1 Ea.
19-103-396 Black 1 Ea.
19-103-397 Brown 1 Ea.
19-103-398 Purple 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 19-103-393 Supplier Brady™ Supplier No. 123246

Includes

1 key

Key retaining safety feature prevents key from being removed until closed

  • Key-retaining steel padlock has a 3 in. reinforced laminated steel shackle
  • Keyed different
  • Hardened steel body withstands severe physical abuse and tampering
  • Offers a lighterweight option in comparison to standard laminated steel padlocks
  • Available in a variety of colors

Specifications

Color Blue
Quantity 1 Ea.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.