Learn More
Brady™ Keyed-Different Aluminum Padlocks with 1 in. Chrome-Plated Steel Shackle Locks
Catalog No. 19103368 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
Yellow
Quantity:
1 Ea.
6/Pk.
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
19-103-368 Purple 6/Pk.
19-103-582 Blue 1 Ea.
19-103-583 Green 1 Ea.
19-103-584 Yellow 1 Ea.
19-103-563 Red 6/Pk.
19-103-367 Purple 1 Ea.
19-103-370 Brown 6/Pk.
19-103-369 Brown 1 Ea.
19-103-564 Blue 6/Pk.
19-103-565 Green 6/Pk.
19-103-566 Yellow 6/Pk.
19-103-567 Orange 6/Pk.
19-103-568 Black 6/Pk.
19-103-581 Red 1 Ea.
19-103-585 Orange 1 Ea.
19-103-586 Black 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 19-103-368 Supplier Brady™ Supplier No. 104573
Includes
2 keys
Lightweight and rugged padlock with solid aluminum body
  • Anodized aluminum finish gives better fade resistance when used outdoors
  • Feature 6-pin cylinders for better tamper resistance
  • Reserved paracentric keyway provides optimal security (prevents someone from getting a copy made at a local key shop)
  • Lightweight, rugged and chemical and corrosion resistant
  • Spark and corrosion resistance
  • Shackle diameter is 1/4 in.
  • Lock body is 1 3/5H x 1 1/2W x 3/4 in. D
  • Lock labels sold separately
  • LOTO-10
Specifications
Color Purple
Quantity 6/Pk.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.