Learn More
Brady™ Keyed-Different Aluminum Padlocks with 3 in. Shackle Locks
Catalog No. 19103434 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
Silver
Yellow
Quantity:
1 Ea.
6/Pk.
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
19-103-434 Green 1 Ea.
19-103-433 Blue 1 Ea.
19-103-436 Orange 1 Ea.
19-103-435 Yellow 1 Ea.
19-103-437 Black 1 Ea.
19-103-438 Brown 1 Ea.
19-103-439 Purple 1 Ea.
19-103-440 Silver 1 Ea.
19-002-114 Black 6/Pk.
19-103-430 Purple 6/Pk.
19-103-429 Orange 6/Pk.
19-103-427 Red 6/Pk.
19-103-431 Silver 6/Pk.
19-002-117 Green 6/Pk.
19-002-116 Brown 6/Pk.
19-002-115 Blue 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 19-103-434 Supplier Brady™ Supplier No. 123295
Add to Cart
Add to Cart
Includes
2 keys
Lightweight padlock with rugged, solid aluminum body
  • Aluminum, keyed-different padlock offers a 3 in. shackle
  • Anodized aluminum finish on the body provides better fade resistance outdoors and a better corrosion resistance in comparison to a standard steel shackle
  • Lightweight yet rugged
  • Features an FDA-approved lubricant in the cylinder and key retaining safety feature
  • Availble in different colors
  • One padlock per pack
Specifications
Color Green
Key Type Keyed Different
Includes Shackle Lock and 2 Keys
Material Aluminum
Quantity 1 Ea.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.