Learn More
Brady™ Keyed-Different Nylon Safety Padlocks with 1.5 in. Shackle Locks
Catalog No. 19103447 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
White
Yellow
Quantity:
1 Ea.
6/Pk.
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
19-103-447 Orange 1 Ea.
19-103-443 Red 1 Ea.
19-103-446 Yellow 1 Ea.
19-103-451 White 1 Ea.
19-103-473 Yellow 6/Pk.
19-103-474 Orange 6/Pk.
19-103-478 White 6/Pk.
19-103-444 Blue 1 Ea.
19-103-445 Green 1 Ea.
19-103-448 Black 1 Ea.
19-103-449 Brown 1 Ea.
19-103-450 Purple 1 Ea.
19-103-470 Red 6/Pk.
19-103-471 Blue 6/Pk.
19-103-472 Green 6/Pk.
19-103-475 Black 6/Pk.
19-103-476 Brown 6/Pk.
19-103-477 Purple 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 19-103-447 Supplier Brady™ Supplier No. 123328
Padlock offers unique keyway which breaks any current traveling form padlock shackle to cylinder
  • Keyed-different padlock with non-conductive, spark resistant lock body
  • Compact and lightweight
  • Available in a variety of colors
Specifications
Color Orange
Key Type Keyed Different
Material Nylon
Quantity 1 Ea.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.