Description
Includes
2 keys
- Made from reinforced laminated steel to withstand severe physical abuse and come with a hardened steel shackle making them extremely difficult to cut
- Body plates are individually coated for superior rust protection
- 5-pin cylinder offers more unique key cuts and better tamper resistance than conventional 4-pin locks
- Lock body size is 1 1/3H x 1 9/16W x 7/8 in. D
- Colored bumpers can be used for identification and protection a
- Lock labels sold separately
Specifications
Specifications
| Color | Purple |
| Quantity | 6/Pk. |
Safety and Handling
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.