Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Lockout Compliance Manual

Lockout Compliance Manual is a binder that keeps your Hazardous Energy Control Procedures organized and protected.

$175.00 - $627.00

Specifications

Format Manual
For Use With (Application) Step-by-step instructions for developing a hazardous energy control program compliance
Subject Safety and Training Materials
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
18-999-1329
View Documents
Brady™
65557
Lockout Binder Vol II
Each for $175.00
Only null left
Add to Cart
 
19-016497
View Documents
Brady™
65558
Lockout Compliance Manual
Each for $627.00
Only null left
Add to Cart
 
Description

Description

  • RoHS compatibility Pass 2011/65/EU
Specifications

Specifications

Manual
Step-by-step instructions for developing a hazardous energy control program compliance
Safety and Training Materials
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.