Learn More
Brady™ Long Body Keyed Alike Safety Padlocks with 1.5 in. Shackle Locks
Catalog No. 19103505 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
White
Yellow
Quantity:
3/Pk.
6/Pk.
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
19-103-505 White 3/Pk.
19-103-497 Red 3/Pk.
19-103-498 Blue 3/Pk.
19-103-499 Green 3/Pk.
19-103-500 Yellow 3/Pk.
19-103-501 Orange 3/Pk.
19-103-502 Black 3/Pk.
19-103-503 Brown 3/Pk.
19-103-504 Purple 3/Pk.
19-103-506 Red 6/Pk.
19-103-507 Blue 6/Pk.
19-103-508 Green 6/Pk.
19-103-509 Yellow 6/Pk.
19-103-510 Orange 6/Pk.
19-103-511 Black 6/Pk.
19-103-512 Brown 6/Pk.
19-103-513 Purple 6/Pk.
19-103-514 White 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 19-103-505 Supplier Brady™ Supplier No. 123422
Padlock offers unique keyway which breaks electrical current from passing padlock shackle to cylinder
  • Lock with 7.6cm (3 in.) long body provides ample space for identification information
  • Made of nylon plastic
  • Lock labels are supplied with English and Spanish or English and French “DO NOT OPERATE” legends
  • Custom keying and engraving is available
  • Available in a variety of colors and packaging
Specifications
Color White
Key Type Keyed Alike
Material Nylon
Quantity 3/Pk.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.