Learn More
Brady™ Long Body Keyed-Different Safety Padlocks with 1.5 in. Shackle Locks
Catalog No. 19103481 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
White
Yellow
Quantity:
1 Ea.
6/Pk.
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
19-103-481 Green 6/Pk.
19-103-491 Yellow 1 Ea.
19-103-479 Red 6/Pk.
19-103-480 Blue 6/Pk.
19-103-482 Yellow 6/Pk.
19-103-483 Orange 6/Pk.
19-103-484 Black 6/Pk.
19-103-485 Brown 6/Pk.
19-103-486 Purple 6/Pk.
19-103-487 White 6/Pk.
19-103-488 Red 1 Ea.
19-103-489 Blue 1 Ea.
19-103-490 Green 1 Ea.
19-103-492 Orange 1 Ea.
19-103-493 Black 1 Ea.
19-103-494 Brown 1 Ea.
19-103-495 Purple 1 Ea.
19-103-496 White 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 19-103-481 Supplier Brady™ Supplier No. 123398
Padlock offers unique keyway which breaks electrical current from passing padlock shackle to cylinder
  • Lock with 7.6cm (3 in.) long body provides ample space for identification information
  • Made of nylon plastic
  • Lock labels are supplied with English and Spanish or English and French “DO NOT OPERATE” legends
  • Custom keying and engraving is available
  • Available in a variety of colors and packaging
Specifications
Color Green
Key Type Keyed Different
Material Nylon
Quantity 6/Pk.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.