Learn More
Brady™ Mini Wall Lock Box
Catalog No. 19000264 Shop All Brady Corporation Products
Change view
Click to view available options
:
Mini Wall Lock Box
Catalog No.
19-000-264 Mini Wall Lock Box
1 options
Catalog No. 19-000-264
Supplier: Brady™ 50938
Mini Wall Lock Box requires fewer locks and is lighter than hasps on delicate energy isolating devices, like circuit breakers.
  • Patented lock rails accommodate up to 8 padlocks and the slot in front allows keys to be inserted when box is locked
  • Use one lock on a each energy control point and place the keys in the lockout box; each worker then places his own lock on the box to prevent access
  • Each employee retains exclusive control, as required by OSHA, by placing his own lock on the lockout box containing the keys to the job locks
  • As long as any one worker's lock remains on the lockout box, the keys to the job locks contained inside cannot be accessed
Specifications
Certifications/Compliance OSHA 1910.147
Dimensions (D x W x H) 5.5 x 4 x 6.2 in.
Width (Metric) 101.6 mm