Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ MiniMark™Accessories

Catalog No. 19018403 Shop All Brady Corporation Products
Change view
Click to view available options
Product Type:
General Industry Starter Package
Pipe Marker Starter Package
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-018-403 General Industry Starter Package
19-018-404 Pipe Marker Starter Package
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-018-403 Supplier Brady™ Supplier No. 105089

MiniMark™ general industrial supply kit for the MiniMark™ Label Printer from Brady™ includes the labels and ribbons you need to get started on your pipe marking labeling projects.

  • The MiniMark™ Starter package includes Markware™ Software Kit with a CD-based software for creating custom safety signs and labels including pipe markers, ANSI signs, custom barcodes, lockout tags.
  • Includes a variety of Indoor/Outdoor vinyl tape and ribbons.
  • Applications: Arc Flash Labeling, Barcode Labeling, Data Communications Labeling, Facility, Safety and Maintenance Identification, Laboratory Identification, Lean/5S Labeling, Lockout Tagout Labeling, Panel Identification, Pipe and Valve Marking, Product Identification, Security Identification

Specifications

Product Type General Industry Starter Package
Color Yellow
For Use With (Equipment) MiniMark Printer
Sterility Non-sterile
Includes MarkWare™ software kit, Vinyl tapes (1 white, 1 yellow of 4 in. x 110 ft., 2.2 in. x 110 ft., 1.12 in. x 110 ft.), 2 white ribbons
Certifications/Compliance UL
For Use With (Application) Arc Flash labeling, Barcode labeling, Data communications labeling, Facility, Safety and maintenance identification, Laboratory identification, Lean/5S labeling, , Lockout tagout labeling, Panel identification, Pipe and valve marking, Product identification, Security identification
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.