Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ MRO Plus Universal All-Purpose Heavy Absorbent Pad

Catalog No. 17111010 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
40 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-010 40 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-010 Supplier Brady™ Supplier No. MRO292P

MRO Plus Universal All-Purpose Heavy Absorbent Pad is designed for cleaning of oil, water-based and chemical liquids.

  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluidsHeavy weight - specifically designed for continuous leaks and larger spills so you use less material
  • 3-ply Meltblown Polypropylene construction is designed for general industrial applications
  • Perforations provide cost control and reduce waste by allowing you to use only what you need

Specifications

Capacity (English) 40 gal.
Capacity (Metric) 151.4 L
Type MRO Plus Universal All-Purpose Heavy Absorbent Pad
Weight Capacity Heavyweight
Length (Metric) 91.44 m
Width (Metric) 76.2 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.