Learn More
Description
- Ideal for outdoor use in waste troughs, coolant tanks, fluid reservoirs and sump wells
- Easy to retrieve and can soak up liquids in hard-to-reach areas
Specifications
Specifications
| Color | White |
| Capacity (English) | 14 gal. |
| Type | Absorbent Pillow |
| For Use With (Application) | Waste troughs, coolant tanks, fluid reservoirs, and sump wells |
| Packaging Quantity | 8/Cs. |
| Dimensions (L x W x H) | 5.08 x 45.75 x 45.72 cm |
| Length (Metric) | 45.72 cm |
| Width (Metric) | 45.72 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.