Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ PowerMark™ Printer Accessories

Catalog No. 19041096 Shop All Brady Corporation Products
Change view
Click to view available options
Product Type:
PowerMark Replacement Wiper Pad
PowerMark Spare Ribbon Cassette
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-041-096 PowerMark Spare Ribbon Cassette
19-041-100 PowerMark Replacement Wiper Pad
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-041-096 Supplier Brady™ Supplier No. 13507

Replacement parts for the PowerMark™ Industrial Sign and Label Printer.

  • Spare ribbon cassette to hold printer ribbon in a clear assembly
  • Replacement wiper pad to keep the printer running clean without smudges

Specifications

Certifications/Compliance RoHS
Color Clear
Format Ribbon
Product Type PowerMark Spare Ribbon Cassette
Sterility Non-sterile
For Use With (Equipment) PowerMark™ printer
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.