Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Production Status Tags

Catalog No. 19040521 Shop All Brady Corporation Products
Change view
Click to view available options
Displayed Text:
DESCRIPTION/PART NO./SERIAL NO./JOB NO./P.O. NO.
Defective Do Not Use Signed By/Date
Equipment Out of Service W.S. No./Signed/Date
Hazardous Waste Description/Source/Liquid/Solid/Weigth/Volume/Hazard(S)/Toxic/Corrosive/Ignitable/Reactive/By/Date
Hoist Inspected O.K. For Use Signed By/Date
Hold Date/By/Reason/Released By/Date
Non-conforming Part No./Serial No./Part Name/Customer/P.O. No. Date/No. Pieces/Reason/Inspector/Date
O.K. To Use Job NO./P.O. No./Description/Quantity/By/Date
O.K. to Ship
Out of Service
Out of Service Date/Ser #/Location/Problem/Signed By
Q.A. Approved W.O. No./Description/Quantity/Comments/By/Date
Ready For Service
Rejected Job No./P.O. No./Part No./Serial No./Part Name/Number Of Pieces Rejected/Reason/Disposition/Inspector/Date
Repair
Repaired Equipment I.D./Description/Job No./Serial No./Condition/Comments/By/Date
Sample
Scrap
Work Order W.O. No./Description/Quantity/Comments/By/Date
Color:
Black on Green
Black on Red
Black on Yellow
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text Color
19-040-521 Hoist Inspected O.K. For Use Signed By/Date Black on Yellow
19-040-538 Non-conforming Part No./Serial No./Part Name/Customer/P.O. No. Date/No. Pieces/Reason/Inspector/Date Black on Yellow
19-040-501 Work Order W.O. No./Description/Quantity/Comments/By/Date Black on Yellow
19-040-499 DESCRIPTION/PART NO./SERIAL NO./JOB NO./P.O. NO. Black on Yellow
19-040-504 Ready For Service Black on Green
19-040-506 Hold Date/By/Reason/Released By/Date Black on Yellow
19-040-510 Out of Service Black on Red
19-040-518 Sample Black on Yellow
19-040-517 Scrap Black on Yellow
19-040-516 Out of Service Date/Ser #/Location/Problem/Signed By Black on Red
19-040-509 Defective Do Not Use Signed By/Date Black on Red
19-040-515 Equipment Out of Service W.S. No./Signed/Date Black on Red
19-040-520 O.K. To Use Job NO./P.O. No./Description/Quantity/By/Date Black on Green
19-040-540 Rejected Job No./P.O. No./Part No./Serial No./Part Name/Number Of Pieces Rejected/Reason/Disposition/Inspector/Date Black on Red
19-040-522 Hazardous Waste Description/Source/Liquid/Solid/Weigth/Volume/Hazard(S)/Toxic/Corrosive/Ignitable/Reactive/By/Date Black on Yellow
19-040-852 Q.A. Approved W.O. No./Description/Quantity/Comments/By/Date Black on Green
19-040-853 O.K. to Ship Black on Yellow
19-040-854 Repair Black on Yellow
19-040-856 Repaired Equipment I.D./Description/Job No./Serial No./Condition/Comments/By/Date Black on Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Catalog No. 19-040-521 Supplier Brady™ Supplier No. 86762
Add to Cart
Add to Cart

Production Status Tags help you effectively monitor and control production and equipment maintenance.

  • The excellent write-on surface of B-853 Cardstock is ideal for adding on-the-spot customized notes and other information
  • Paper cardstock tags are recommended for indoor use only
  • Includes 3/8 in. metal grommet
  • Not recommended for chemical exposure

Specifications

Product Type Production Status Tag
Color Black on Yellow
Material Cardstock
Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
Diameter (English) Shackle 0.375 in.
Diameter (Metric) Shackle 9.52 mm
Dimensions (L x W) 3 x 5.75 in.
Displayed Text Hoist Inspected O.K. For Use Signed By/Date
For Use With (Application) To help you monitor and control production and equipment maintenance
Height (English) 3 in.
Height (Metric) 76.2 mm
Includes (100) 12 in. cotton string ties
Temperature Range (English) −40° to +194°F
Temperature Range (Metric) −40° to +90°C
Thickness 0.25 mm
Width (English) 5.75 in.
Width (Metric) 146 mm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.