Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Quick-Align™ Symbols

Catalog No. 191302557 Shop All Brady Corporation Products
Change view
Click to view available options
Displayed Text:
ACID
ALK
COR
No Water
OXY
Height (English):
1 in.
2 in.
3 in.
4 in.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text Height (English)
19-130-2557 No Water 1 in.
19-130-2578 ACID 4 in.
19-130-2577 OXY 4 in.
19-130-2580 ALK 4 in.
19-130-2579 COR 4 in.
19-130-2565 No Water 2 in.
19-130-2571 No Water 3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-130-2557 Supplier Brady™ Supplier No. 58505

For use with Blank Hazardous Communication Panels to identify hazardous materials in accordance with the NFPA system

  • Tough outdoor-grade self-sticking vinyl characters are black Gothic style (except for radiation trefoil)
  • Speciific hazard warnings (“Use no water”, radiation symbol, oxidizer, acid, alkalai, corrosive)

Always consult Material Safety Data Sheets for product-specific hazard information.

Order Info

Each package contains 10 identical characters.

Specifications

Certifications/Compliance RoHS
Material Vinyl
Quantity 10/Pk.
Style Gothic
For Use With (Application) Indoor and outdoor use, used for pipemarkers, arrow tape, numbers and letters, vehicle placards, pedestal markers, warning panels and safety signs
Height (English) 1 in.
Max. Temperature (English) 180°F
Min. Temperature (English) -40°F
Thickness 0.004 in.
Temperature Range (Metric) -40°C to 82°C
Temperature Range (English) -40°F to 180°F
Displayed Text No Water
Product Type Hazardous Material Identifier
Color Black
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.