Learn More
Description
- Tough outdoor-grade self-sticking vinyl characters are black Gothic style (except for radiation trefoil)
- Speciific hazard warnings (“Use no water”, radiation symbol, oxidizer, acid, alkalai, corrosive)
Always consult Material Safety Data Sheets for product-specific hazard information.
Order Info
Each package contains 10 identical characters.
Specifications
Specifications
| Certifications/Compliance | RoHS |
| Material | Vinyl |
| Quantity | 10/Pk. |
| Style | Gothic |
| For Use With (Application) | Indoor and outdoor use, used for pipemarkers, arrow tape, numbers and letters, vehicle placards, pedestal markers, warning panels and safety signs |
| Height (English) | 2 in. |
| Max. Temperature (English) | 180°F |
| Min. Temperature (English) | -40°F |
| Thickness | 0.004 in. |
| Temperature Range (Metric) | -40°C to 82°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.