Description
- Made with Re-Form sorbent material
- Absorbs more than 34 times its weight in liquid
- Spunbound coverstock on both sides for more durability and strength
Specifications
Specifications
| Capacity (English) | 35.9 gal./Bale |
| Type | Oil-only Heavyweight Pad |
| For Use With (Application) | For oil and solvent-based liquids |
| Packaging Quantity | 100/Cs. |