Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Safety First: If You Get Chemicals On Your Body… Sign

Catalog No. 19035744 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
10 x 14 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-035744 10 x 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-035744 Supplier Brady™ Supplier No. 84476

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents. Safety First: If You Get Chemicals On Your Body… Sign B-302 is an high performance pressure sensitive safety sign.

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads “SAFETY FIRST: IF YOU GET CHEMICALS ON YOUR BODY OR EYES, WASH THOROUGHLY WITH PLENTY OF WATER”
  • Green/black on white
  • Size: 25.4H x 35.56Wcm (10H x 14W in.)
  • B-302 self-sticking polyester—surface printed polyester, polyester overlaminate and sensitive acrylic
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

Specifications

Certifications/Compliance Globally Harmonized System of Classification and Labeling of Chemicals (GHS); NFPA 704;OSHA 1910.1200
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Mounting Type Surface Mount
For Use With (Application) Identification of fire alarms, extinguishers and emergency exits
Height (English) 10 in.
Max. Temperature (English) 230°F
Min. Temperature (English) -40°F
Thickness 0.006 to 0.01 in.
Width (English) 14 in.
Width (Metric) 35.56 cm
Temperature Range (Metric) -40°C to 110°C
Temperature Range (English) -40°F to 230°F
Dimensions (L x W) 10 x 14 in.
Displayed Text Safety First If You Get Chemicals On Your Body or Eyes Wash Thoroughly With Plenty of Water
Product Type Chemical and Hazardous Materials Sign
Color Black
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.