Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Self-Laminating Tags

Catalog No. 19040549 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black, Red, White
Black, Yellow
Yellow
Message:
CAUTION Do Not Remove This Tag
CAUTION Do Not Remove This Tag/Remarks
DANGER DO NOT OPERATE Do Not Remove This Tag It is Here To Protect My Life. Name/Date Do Not Remove This Tag/Remarks
DANGER DO NOT OPERATE Until this tag is removed by:/Signed By/Date/Phone Do Not Remove This Tag/Remarks
DANGER DO NOT OPERATE/ Signed By/Date/Phone Do Not Remove This Tag/Remarks
DANGER Do Not Remove This Tag/Remarks/See Other Side
DANGER Out Of Service/Signed By/Date/Phone Do Not Remove This Tag/Remarks
Dimensions (L x W):
5.75 x 3 in.
7 x 4 in.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Message Dimensions (L x W)
19-040-549 Yellow CAUTION Do Not Remove This Tag/Remarks 7 x 4 in.
19-016467 Black, Red, White DANGER DO NOT OPERATE Do Not Remove This Tag It is Here To Protect My Life. Name/Date Do Not Remove This Tag/Remarks 5.75 x 3 in.
19-040-548 Black, Red, White DANGER Do Not Remove This Tag/Remarks/See Other Side 7 x 4 in.
19-040-542 Black, Yellow CAUTION Do Not Remove This Tag 5.75 x 3 in.
19-040-551 Black, Red, White DANGER DO NOT OPERATE/ Signed By/Date/Phone Do Not Remove This Tag/Remarks 7 x 4 in.
19-040-552 Black, Red, White DANGER Out Of Service/Signed By/Date/Phone Do Not Remove This Tag/Remarks 7 x 4 in.
19-040-804 Black, Red, White DANGER DO NOT OPERATE Until this tag is removed by:/Signed By/Date/Phone Do Not Remove This Tag/Remarks 5.75 x 3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-040-549 Supplier Brady™ Supplier No. 86514
Add to Cart
Add to Cart

Includes

10 Nylon ties

Self-Laminating Tags can be personalized by writing on special instructions, hazards or warnings.

  • B-851 Rigid Economy Polyester with brass grommets, exceeds OSHA's 50 lb pull force requirement for Tagout devices
  • Textured write-on surface allows for authorized signatures, dates and other special instructions
  • Seal and protect your written message with a clear adhesive overlay
  • Withstands moisture, grease and dirt

Specifications

Quantity 10/Pk.
Certifications/Compliance OSHA
Color Yellow
Diameter (English) Grommet 0.37 in.
Material Polyester
Message CAUTION Do Not Remove This Tag/Remarks
For Use With (Application) For marking on-the-spot warning of dangerous conditions
Durability 3 to 5 Years
Height (English) 7 in.
Height (Metric) 17.7cm
Thickness 0.01 in.
Temperature Range (Metric) -40°C to 100°C
Temperature Range (English) -40°F to 212°F
Width (English) 4 in.
Width (Metric) 10.1 cm
Dimensions (L x W) 7 x 4 in.
Diameter (Metric) Hole 0.93 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.