Learn More
Brady™ Signs: DANGER: ASBESTOS CANCER AND LUNG DISEASE HAZARD DO NOT DISTURB WITHOUT PROPER TRAINING AND EQUIPMENT
Shop All Brady Corporation ProductsDescription
- Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
- Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
- Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
- Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)
Specifications
Specifications
| Certifications/Compliance | Globally Harmonized System of Classification and Labeling of Chemicals (GHS);NFPA 704;OSHA 1910.1200 |
| Material | Polyester |
| Adhesive Type | Permanent Acrylic |
| Height (English) | 14 in. |
| Thickness | 0.01 in. |
| Width (Metric) | 25.4 cm |
| Dimensions (L x W) | 14 x 10 in. |
| Color | Yellow on White |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
