Learn More
Brady™ TLS2200 and TLS-PC Link™Portable Thermal Transfer Nylon Printer Labels for 0.5 to 8mL Vials/Tubes
Labels for 0.5 to 8mL vials or tubes
Specifications
Certifications/Compliance Halogen-free (DIN VDE 0472/parts 815), UL
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Format Label
View More Specs
Products 2
Catalog Number Mfr. No. Height (English) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Width (English) Price Quantity & Availability  
11-877-56
View Documents
Brady™
PTL84499
0.5 in. 0.9 in. N/A N/A N/A
11-877-43
View Documents
Brady™
PTL11499
0.75 in. 0.5 in. N/A N/A N/A
Description
Description
  • Durable, chemical- and solvent-resistant labels; ink will not smear or fade, and the text stays crisp and readable
  • Printable labels allow more information to fit on any size legend
  • Ideal for different environments from liquid nitrogen to autoclaves
Order Info
Labels available in other materials and sizes; contact your Customer Service Representative for ordering information.
Specifications
Specifications
Halogen-free (DIN VDE 0472/parts 815), UL
White
Label
TLS 2200™/TLS-PC Link™ Label
1
Rectangle
-196°C to +90°C
Acrylic, Permanent
Matte
Nylon Cloth
BMP™71, TLS 2200™, TLS-PC Link™
500/Pk.
-320 to 194°F
0.165 mm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.