Learn More
Brady™ Vehicle Placards
Catalog No. 19807139 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Silver
White on Green
White on Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-807-139 White on Green
19-017184 Silver
19-807-140 White on Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-807-139 Supplier Brady™ Supplier No. 63407
Add to Cart
Add to Cart
Durable placards for vehicles
Models 63407 and 63408
  • Text says NON-FLAMMABLE GAS 2 (19807139) or FLAMMABLE GAS 2 (19807140)
  • Square corners and no holes
  • Size: 27.3 × 27.3cm (10.75 × 10.75 in.)
  • B-946 self-sticking vinyl — may be applied directly to vehicle; provides more than five years of outdoor durability
Model 76988
  • Front plate holder for use with back plate placard holder Model 76989
  • Size: 27.3 × 27.3cm (10.75 × 10.75 in.)
  • Heavy-duty aluminum
Compliance
Meet D.O.T. requirements HM-181 requirements
Specifications
Certifications/Compliance HM-181
Material Vinyl
Quantity 1 Ea.
Height (English) 10.75 in.
Width (English) 10.75 in.
Displayed Text Non-Flammable Gas 2
Product Type Standard Vehicle Placard
Color White on Green
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.