Learn More
Description
- Nonconductive; can be safely used in electrically hazardous areas
- Non-rusting (unlike metal posts)
- Hollow base can be filled with sand for more stable application
- Large C-hook holds chain in place on large posts and can be used open or closed for greater security
- Nonreflective
Specifications
Specifications
| Color | Yellow, Magenta |
| Material | Polyethylene, Polystyrene |
| For Use With (Application) | Used in electrically hazardous areas |
| Height (Metric) | 104.14 cm |
| Type | Non-conductive Warning Post |
| Diameter (Metric) Outer | 7.62 cm |
| Diameter (Metric) Base | 40.64 cm |
| Diameter (English) Outer | 3 in. |
| Height (English) | 41 in. |
| Diameter (English) Base | 16 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.