Learn More
Brady™ BradyLam™ Fiberglass with Polyester Laminate Sign: PESTICIDE SPRAYING
Catalog No. 19106768 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
12 x 9 in.
Catalog No. Dimensions (L x W)
19-106-768 12 x 9 in.
1 options
Catalog No. 19-106-768 Supplier Brady™ Supplier No. 17535
Improve workplace safety with proper signage and markings. This Brady™ laminated fiberglass sign is a durable, clear and effective tool to raise awareness to workplace hazards.
  • Fiberglass material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Fiberglass out-performs all other sign materials in terms of strength, durability, and longevity
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Material Fiberglass, Polyester
Shape Rectangular
Dimensions (L x W) 12 x 9 in.
Displayed Text PESTICIDE SPRAYING
Color Red on White
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.