Learn More
Description
- Convenient carryall can be used as a full on-the-go print station
- Includes compartments sized to fit your phone, materials and cords so you can keep everything you need together
- Has an adjustable padded shoulder strap so you can keep working comfortably
- Soft cover protects the printer during transportConvenient carryall can be used as a full on-the-go print station
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | BradyPrinter M611 |
| Material | Vinyl |
| Dimensions (L x W x H) | 4.75 x 30 x 19 in. |
| Quantity | 1 Ea. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.