Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ BradyPrinter S3100 Sign and Label Printer and Lockout Tagout Materials Kit for LINK360™ Software
Get more out of your LINK360™ Safety Software when you add the power of on-demand printing with the BradyPrinter S3100 Sign and Label Printer and Lockout Tagout Materials Kit for LINK360™ Software.
Supplier: Brady™ S3100WOLINK360
Description
- Custom-curated kit of printer, ribbons, labels and tags has everything you need to add tag and label printing capabilities to your LINK360™ Safety Software solution
- S3100 Printer offers a seamless experience and high versatility
- Simple, durable, reliable printer can print energy source labels and lockout tags from LINK360™ plus a wide range of labels and signs thanks to onboard editing that allows workers to walk up, create and print
- Smear-proof B30 Series R10000 Ribbons (black, red) produce multiyear-durable printed text for industrial areas and outdoor conditions
- OSHA DANGER LOTO Tags combine a printable side with textured write-on surface for authorized signatures, dates or special instructions
- Blank Valve Tags and B30 Series Continuous Indoor Outdoor Vinyl Labels let you easily create and apply energy source tags
- LINK360™ Safety Software sold separately
Specifications
Label Printer Kit | |
Color LCD Touchscreen | |
Plastic | |
10.25 in. | |
260.35 mm | |
9.5 in. | |
10.25 x 12 x 9.5 in. | |
Non-sterile | |
1024 x 800 px | |
Chinese (Simplified), Chinese (Traditional), Dutch, English, French, German, Hindi, Italian, Japanese, Portuguese, Russian, Spanish, Thai, Vietnamese, Czech | |
S3100 | |
300 dpi | |
Ethernet, USB | |
-40°F to +180°F | |
UL, cUL, CE, WEEE, RoHS, RCM (ANZ); EAC (Russia, Belarus, Kazakhstan); CCC-RoHS (China) | |
5 Years | |
110/240 V |
Blue, Gray | |
Brady Labels | |
Thermal Transfer | |
12 in. | |
304.8 mm | |
241.3 mm | |
Not Autoclavable | |
Single Color | |
LINK360 Starter Bundle, S3100 Sign and Label WIFI Printer, 3.25 x 5.75 in. OSHA DANGER Polyester Tags, 4.33 in. x 200 ft. R10000 Black Printer Ribbon, 3 in. x 100 ft. White Outdoor Vinyl, 4.33 in. x 200 ft. R10000 Red Printer Ribbon, 10 Blank White Valve Tags (1.5 x 3 in.) | |
256 MB | |
3 in./sec. | |
Brady Workstation, Brady LINK360, Windows-based Driver for 3rd Party Software Use | |
Windows 7, Windows 8, Windows 10, Windows 11 | |
10°C to 40°C | |
Hazard Communication Identification, Barcode Labeling, Lockout Tagout, Pipe and Valve Marking | |
AC Adaptor | |
QWERTY (US) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction