Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BradyPrinter S3700 Multicolor and Cut Sign and Label Printer and Pipe Marker Label Materials Kit

Catalog No. 17377003 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Blue, Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-377-003 Blue, Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-377-003 Supplier Brady™ Supplier No. S3700KITPIPE
Add to Cart
Add to Cart
Request a Quote

Pipe marking projects have never been easier thanks to this convenient starter kit, complete with the ANSI standard colors you need and a smooth print-on-demand solution.

  • Printer optimized for 1-, 2- or 4-color panel ribbons for multicolor made easy
  • No calibrating, no threading, no waste (single-color ribbon only in kit)
  • Create shapes, arrows, letters and more with the built-in XY plotter cutter that slices straight lines, smoother curves and tighter connection points
  • Get your print jobs done right in less time with 20-second ribbon and material changeovers plus up to 5 in./sec. print speed
  • Empower any user to print how they choose with time-saving onboard templates, intuitive PC labeling software and a mobile app
  • Simply tap, print, go with preprogrammed onboard label templates plus 600 barcodes and symbols
  • Durable, low-shrink All Weather Permanent Adhesive Vinyl Label Tape with ultra-aggressive adhesive is ideal for labeling pipes indoors and out
  • Waste less, print more with print ribbons that last up to 60% longer by operating independently of label roll

Specifications

Product Type Label Printer Kit
Color Blue, Gray
Display Type Color LCD Touchscreen
For Use With (Equipment) Brady Labels
Material Plastic
Print Technology Thermal Transfer
Length (English) 14.25 in.
Width (English) 19 in.
Length (Metric) 361.95 mm
Width (Metric) 482.6 mm
Height (English) 11 in.
Height (Metric) 279.4 mm
Dimensions (L x W x H) 14.25 x 19 x 11 in.
Autoclavable Not Autoclavable
Sterility Non-sterile
Color Capability Multicolor
Includes S3700 Safety and Facility Printer, Brady Workstation Safety and Facility ID Software, Cleaning Kit, Mouse, Power Cord, Quick Start Guide, USB Cord, USB Flash Drive, 4 in. Continuous Yellow, Blue, White, Red, and Green Vinyl Labels, 2.25 in. Green, Red, and Blue Continuous Vinyl Labels, Black Ribbon Cartridge, 3 White Ribbon Cartridges
Languages Indonesian, Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese, Romanian, Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese
Memory 512 MB
Model S3700
Print Speed 5 in./sec.
Resolution 300 dpi
Temperature Range (English) 59°F to 104°F
Temperature Range (Metric) 15°C to 40°C
For Use With (Application) Asset Tracking, Cold Temperature Labeling, Equipment Labeling, Electrical Labeling, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Food and Beverage Process Labeling, Healthcare and Hospital Labeling, Barcode Labeling
Warranty 5 Years
Power Supply Details AC Adaptor
Voltage 110/240 V
Keyboard Type QWERTY (US)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.