Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BradySnap-On™ Pipe Markers, Legend: (BLANK)

Catalog No. 19104716 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black, Yellow
White, Blue
White, Green
White, Red
Description:
Style A2L
Style B2L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Description
19-104-716 Black, Yellow Style B2L
19-104-713 White, Green Style A2L
19-104-714 White, Red Style A2L
19-104-718 White, Red Style B2L
19-104-719 White, Blue Style A2L
19-104-717 White, Green Style B2L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-104-716 Supplier Brady™ Supplier No. 42304
Add to Cart
Add to Cart

Installs quickly and easily in older facilities or new construction. Brady BradySnap-On Pipe Marker, Legend: (BLANK) is made of coiled printed plastic sheet (B-915) and ideal for indoor and outdoor use.

  • Precoiled to snap around pipe
  • Ideal for indoor and outdoor use
  • Applied to dirty, oily, greasy or rough surfaces

Specifications

Certifications/Compliance ASME (ANSI)
Description Style B2L
Color Black, Yellow
Material Plastic (B-915)
Product Type Pipe Marker
For Use With (Application) Pipe and valve marking
Warranty 5 to 8 Years
Diameter (English) 1.125 to 2.375 in.
Diameter (Metric) 28.6 to 60.3 mm
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.