Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human BRAF [V599E], GST Tag Recombinant Protein

Catalog No. PR7304A
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR7304A 100 μg
PV4173 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR7304A Supplier Invitrogen™ Supplier No. PR7304A
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic

BRAF acts upstream of MEK in the MAP kinase pathway and is involved in cell division, differentiation, and secretion. The BRAF mutant V599E is oncogenic and found in a variety of cancers. Mutations in BRAF (V599E Mutant) have also been associated with various cancers, including non-Hodgkin lymphoma, colorectal cancer, malignant melanoma, thyroid carcinoma, non-small cell lung carcinoma, and adenocarcinoma of lung. A pseudogene, which is located on chromosome X, has been identified for BRAF (V599E Mutant).
TRUSTED_SUSTAINABILITY

Specifications

Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Molecular Weight (g/mol) 67.1 kDa
Name Human BRAF [V599E], GST Tag
Quantity 100 μg
Regulatory Status RUO - research use only
Common Name BRAF (V599E Mutant)
Kinase Family RAF Kinase Family
Protein Length 416-766; V599E
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.