Learn More
Cellular Engineering Technologies Yeast-Expressed Human brain-derived neurotrophic factor (BDNF). Vial of lyophilized power (100µg)
Supplier:  Cellular Engineering Technologies GR1001100C
Encompass_Preferred
Human Brain-Derived Neurotrophic Factor (BDNF) is an activity-dependent neurotrophic factor that supports neurons' differentiation, maturation, and survival3 and provides a neuroprotective effect under adverse conditions. BDNF stimulates and controls the growth of new neurons from neural stem cells.
Catalog No. NC2753128
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.