Learn More
Teknova Brain Heart Infusion Agar Plates with Kanamycin-25. 100mm Plates, Sterile. 20 Pack 8 pack.
SDP
Supplier:  Teknova B10978PK
Encompass_Preferred
3.7% Brain Heart Infusion,1.5% Agar,25µg/mL Kanamycin Sulfate
Catalog No. 50-238-7736
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.