Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Brain Punch Set
SDP

Catalog No. 10000245 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
0.25 to 1.25 mm
1.5 to 2 mm
Includes:
Set of 3
Set of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Includes
10-000-245 0.25 to 1.25 mm Set of 5
10-000-246 1.5 to 2 mm Set of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-245 Supplier Stoelting™ Supplier No. 57401

Stoelting™ Brain Punch Set features spring loaded nylon expellers and a convenient handle

  • For the microdissection of frozen brain sections
  • Facilitates the Palkovits Punch technique of dissecting out specific brain nuclei for neurochemistry.
  • Nuclei are punched from frozen brain sections on glass slides, and expelled into the reagents.
  • Spring-loaded expellers avoid risk of contamination that might be caused by expelling with breath
  • Each punch has an electropolished sharp end for smooth cuts and minimal tissue adhesion
  • Smaller tubes are reinforced by an outer layer of concentric tubing except at the tip.
  • Slight, ±5%, size variation due to electropolish etching to sharpen
  • Replacement punches available individually

Specifications

For Use With (Application) Neuroscience
Size 0.25 to 1.25 mm
Includes Set of 5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.